Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse FABP1/L-FABP Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP26197950UG

 View more versions of this product

Catalog No. NBP26197950


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-127 of Mouse FABP1/L-FABP The Recombinant Mouse FABP1/L-FABP Protein is derived from E. coli. The Recombinant Mouse FABP1/L-FABP Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25 mg/ml
Phosphate Buffer Saline (pH7.4) containing 20% glycerol, 1mM DTT
16.6 kDa
50 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
FABP1
Mouse
SDS-Page
2168
>95%, by SDS-PAGE
E. coli
RUO
FABP1/L-FABP
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit