Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse IL-11 Animal-Free Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP2762452UG

 View more versions of this product

Catalog No. NBP2762452U


Add to cart

Description

Description

An un-tagged bioactive recombinant protein corresponding to Mouse IL-11

The Recombinant Mouse IL-11 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications

Specifications

Lyoph
3589
>95%, by SDS-PAGE
E. coli
IL11
Sterile water at 0.1 mg/mL
SDS-Page, Bioactivity
19.3 kDa
2 μg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
IL-11 is a monomer, 19.3 kDa (179 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit