Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-17A Protein, CF

New rm IL-17A is now available. It is CHO expressed and has a lower endotoxin specification

$177.50 - $723.50

Specifications

Accession Number Q62386.1
Buffer Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
Conjugate Unconjugated
Endotoxin Level <0.10 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
42-1ML-010CF
View Documents
R&D Systems™
421ML010/CF
10 µg
Each for $177.50
Add to cart
 
42-1ML-100CF
View Documents
R&D Systems™
421ML100/CF
100 µg
Each for $723.50
Add to cart
 
Description

Description

  • Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells.
  • ED50 for this effect is 0.25 to 1.25 ng/mL.
Specifications

Specifications

Q62386.1
Unconjugated
Bioactivity
15.5 kDa (monomer)
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
E. coli-derived mouse IL-17/IL-17A protein Thr22-Ala158
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
<0.10 EU per 1 µg of the protein by the LAL method.
3605
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Reconstitute at 100 µg/mL in sterile 4 mM HCl.
Mouse
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit