Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-21 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

$489.00 - $5200.00

Specifications

Accession Number Q9ES17.1
Conjugate Unconjugated
Cross Reactivity IL-21
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
59-4ML0-25
View Documents
R&D Systems™
594ML025
25 μg
Each for $489.00
Add to cart
 
59-4ML1-00
View Documents
R&D Systems™
594ML100
100 μg
Each for $989.00
Add to cart
 
59-4ML0-1M
View Documents
R&D Systems™
594ML01M
1000 μg
Each for $5,200.00
Add to cart
 
Description

Description

The Recombinant Mouse IL-21 Protein is derived from E. coli. The Recombinant Mouse IL-21 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Q9ES17.1
IL-21
Bioactivity
59067
Mouse
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Reconstitute at 100 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
<EM>E. coli</EM>-derived mouse IL-21 protein Pro25-Ser146, with and without an N-terminal Met
Unconjugated
<0.01 EU per 1 μg of the protein by the LAL method.
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Il21
14.4 kDa
25 μg
RUO
Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit