Learn More
R&D Systems™ Recombinant Mouse IL-22BP His-tag Protein CF Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9597BP050
Catalog No. 9597BP050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Mouse IL-22BP His-tag Protein is derived from NS0. The Recombinant Mouse IL-22BP His-tag Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Q80XF5
Bioactivity
116379
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Mouse myeloma cell line, NS0-derived Thr19-Pro230, with a C-terminal 10-His tag
<0.10EU per 1μg of the protein by the LAL method.
Recombinant Protein
Mouse
Bioactivity
Lyophilized from a 0.2μm filtered solution in PBS.
26kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein,mgC150509,mgC150510, zcytoR16
Unconjugated
Reconstitute at 250μg/mL in PBS.