Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse IL-4 Animal-Free Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

$83.00 - $3732.64

Specifications

Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
Concentration Lyoph
For Use With (Application) SDS-Page, Bioactivity
Gene ID (Entrez) 3565
Gene Symbol IL4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2762705U
View Documents
Novus Biologicals™
NBP2762705UG
5 μg
Each for $83.00
Add to cart
 
NB27627020U
View Documents
Novus Biologicals™
NBP27627020UG
20 μg
Each for $188.00
Add to cart
 
NBP2762701M
View Documents
Novus Biologicals™
NBP2762701MG
1 mg
Each for $3,732.64
Add to cart
 
Description

Description

An un-tagged bioactive recombinant protein corresponding to Mouse IL-4

The Recombinant Mouse IL-4 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications

Specifications

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
SDS-Page, Bioactivity
IL4
>95%, by SDS-PAGE
E. coli
Store at -20 to -80°C. Avoid freeze-thaw cycles.
Lyoph
3565
13.7 kDa
Sterile water at 0.1 mg/mL
IL-4 is a monomer, 13.7 kDa (121 aa)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit