Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse MDH2 Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$191.70 - $494.50
Specifications
Concentration | 1 mg/ml |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate buffer saline (pH7.4) containing 20% glycerol |
Gene ID (Entrez) | 4191 |
Molecular Weight (g/mol) | 35.4 kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP26145020
|
Novus Biologicals™
NBP26145020UG |
20 μg |
Each for $191.70
|
|
|||||
NB261450100
|
Novus Biologicals™
NBP261450100UG |
100 μg |
Each for $494.50
|
|
|||||
Description
A recombinant protein with a N-terminalHis-tag and corresponding to the amino acids 25-338 of Mouse MDH2. The Recombinant Mouse MDH2 Protein is derived from E. coli. The Recombinant Mouse MDH2 Protein has been validated for the following applications: SDS-Page.Specifications
1 mg/ml | |
Phosphate buffer saline (pH7.4) containing 20% glycerol | |
35.4 kDa | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
< 1.0 EU per 1μg of protein (determined by LAL method) | |
MDH2 | |
>95%, by SDS-PAGE |
SDS-PAGE | |
4191 | |
E. coli | |
RUO | |
MDH2 | |
Mouse |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title