Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Park7/DJ-1 Protein

Highly purified. Generating reliable and reproducible results.

$280.50 - $795.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol PARK7
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25952305 View Documents Novus Biologicals™
NBP2595230.5MG
0.5mg Each for $795.00
Add to cart
 
NBP259523X View Documents Novus Biologicals™
NBP259523100UG
100μg Each for $280.50
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-189 of Mouse Park7/DJ-1 The Recombinant Mouse Park7/DJ-1 Protein is derived from E. coli. The Recombinant Mouse Park7/DJ-1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

E.coli
RUO
PARK7
11315
Recombinant Mouse Park7/DJ-1 Protein
Liquid. PBS (pH 7.4) containing 20% glycerol,1mM DTT
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
22.4 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit