Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Pentraxin 2/SAP His-tag Protein, CF
Measured by its binding ability in a functional ELISA.
Supplier: R&D Systems™ 2558SAB050
This item is not returnable.
View return policy
Description
When recombinant Mouse Pentraxin/SAP His-tag (2558-SAB) is immobilized at 10 μg/mL (100 μL/well), the concentration of Biotinylated recombinant human FC gamma RIIA/CD32a (R167) Protein that produces 50% of the optimal binding response is found to be approximately 0.5-2.5 μg/mL.Specifications
P12246 | |
325 | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. | |
Mouse myeloma cell line, NS0-derived mouse Pentraxin 2/SAP protein Gln21-Asp224, with a C-terminal 10-His tag | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Reconstitute at 100 μg/mL in sterile water. |
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl2. | |
Observed: 27-35 kDa, under reducing conditions; Theroretical: 25 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Unconjugated | |
Pentraxin 2/SAP |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title