Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse S100B Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP2530700.5MG

 View more versions of this product

Catalog No. NB25307005


Add to cart

Description

Description

An un-tagged recombinant protein corresponding to the amino acids 1-92 of Mouse S100B The Recombinant Mouse S100B Protein is derived from E. coli. The Recombinant Mouse S100B Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

SDS-Page
6285
Mouse S100B Protein
0.5mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Recombinant Protein
Mouse
Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
10.7kDa
>95%, by PAGE
MSELEKAMVA LIDVFHQYSG REGDKHKLKK SELKELINNE LSHFLEEIKE QEVVDKVMET LDEDGDGECD FQEFMAFVAM VTTACHEFFE HE
RUO
S100B
Unlabeled
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit