Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse SPARC-like 1/SPARCL1 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$345.50 - $1606.00

Specifications

Concentration 0.25 mg/mL
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity >90% SDS-PAGE
Source Baculovirus
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB002935
View Documents
Novus Biologicals™
NBP2774900.1MG
0.1 mg
Each for $524.00
Add to cart
 
NB002934
View Documents
Novus Biologicals™
NBP2774900.05MG
0.05 mg
Each for $345.50
Add to cart
 
NB002936
View Documents
Novus Biologicals™
NBP2774900.5MG
0.5 mg
Each for $1,606.00
Add to cart
 
Description

Description

A recombinant protein with a His-Tag and corresponding to the amino acids 17-650aa of Mouse SPARC-like 1/SPARCL1.

Source: Baculovirus

Amino Acid Sequence:

IPTSTRFLSD HSNPTTATLV TPEDATVPIA GVEATADIEN HPSDKAEKPS ALNSEEETHE QSTEQDKTYS FEVDLKDEED GDGDLSVDPT EGTLTLDLQE GTSEPQQKSL PENGDFPATV STSYVDPNQR ANITKGKESQ EQPVSDSHQQ PNESSKQTQD LKAEESQTQD PDIPNEEEEE EEDEEEEEEE EPEDIGAPSD NQEEGKEPLE EQPTSKWEGN REQSDDTLEE SSQPTQISKT EKHQSEQGNQ GQESDSEAEG EDKAAGSKEH IPHTEQQDQE GKAGLEAIGN QKDTDEKAVS TEPTDAAVVP RSHGGAGDNG GGDDSKHGAG DDYFIPSQEF LEAERMHSLS YYLKYGGGEE TTTGESENRR EAADNQEAKK AESSPNAEPS DEGNSREHSA GSCTNFQCKR GHICKTDPQG KPHCVCQDPE TCPPAKILDQ ACGTDNQTYA SSCHLFATKC RLEGTKKGHQ LQLDYFGACK SIPACTDFEV AQFPLRMRDW LKNILMQLYE PNPKHGGYLN EKQRSKVKKI YLDEKRLLAG HPIELLLRD FKKNYHMYVY PVHWQFNELD QHPADRILTH SELAPLRASL VPMEHCITRF FEECDPNKDK HITLKEWGHC FGIKEEDIDE NLLFLEHHHH HH>

Specifications

Specifications

0.25 mg/mL
< 1.0 EU per 1 μg of protein (determined by LAL method)
>90% SDS-PAGE
Baculovirus
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit