Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse STI1 Protein

Highly purified. Generating reliable and reproducible results.

$280.50 - $783.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol STIP1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP259538X View Documents Novus Biologicals™
NBP259538100UG
100μg Each for $280.50
Add to cart
 
NBP25953805 View Documents Novus Biologicals™
NBP2595380.5MG
0.5mg Each for $783.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-543 of Mouse STI1 The Recombinant Mouse STI1 Protein is derived from E. coli. The Recombinant Mouse STI1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

E.coli
RUO
STIP1
10963
Recombinant Mouse STI1 Protein
Liquid. 20mM Tris-HCl(pH8.0) containing 0.1M NaCl, 1mM DTT, 10% glycerol
Unlabeled
1mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Hop, HOPSTI1L, Hsc70/Hsp90-organizing protein, Hsp70/Hsp90-organizing protein, IEF-SSP-3521, NY-REN-11 antigen, Renal carcinoma antigen NY-REN-11, STI1P60, stress-induced-phosphoprotein 1, Transformation-sensitive protein IEF SSP 3521
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
64.7 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit