Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse UCH-L1/PGP9.5 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$133.00 - $358.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol UCHL1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25958920 View Documents Novus Biologicals™
NBP25958920UG
20μg Each for $133.00
Add to cart
 
NBP259589X View Documents Novus Biologicals™
NBP259589100UG
100μg Each for $358.00
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-223 of Mouse UCH-L1/PGP9.5 The Recombinant Mouse UCH-L1/PGP9.5 Protein is derived from E. coli. The Recombinant Mouse UCH-L1/PGP9.5 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

E.coli
RUO
UCHL1
7345
Recombinant Mouse UCH-L1/PGP9.5 Protein
Liquid. PBS containing 10% glycerol, 1mM DTT
Unlabeled
1mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquit
Recombinant Protein
>90%, by SDS-PAGE
Functional, SDS-Page
27.2 kDa
Mouse

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit