Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse VAMP-7 Protein

Highly purified. Generating reliable and reproducible results.

$280.50 - $783.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol VAMP7
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25957610 View Documents Novus Biologicals™
NBP25957610UG
10μg Each for $280.50
Add to cart
 
NBP25957650 View Documents Novus Biologicals™
NBP25957650UG
50μg Each for $783.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-94 of Mouse VAMP-7 The Recombinant Mouse VAMP-7 Protein is derived from E. coli. The Recombinant Mouse VAMP-7 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

E.coli
RUO
VAMP7
6845
Recombinant Mouse VAMP-7 Protein
Liquid. PBS (pH7.4) containing 1mM EDTA, 0.1mM PMSF, 10% glycerol
Unlabeled
1mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
FLJ54296, SYBL1FLJ53762, synaptobrevin-like 1, Synaptobrevin-like protein 1, tetanus neurotoxin-insensitive VAMP, Tetanus-insensitive VAMP, Ti-VAMP, TIVAMP, VAMP-7FLJ53045, vesicle-associated membrane protein 7
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
12.8 kDa
Mouse

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit