Learn More
R&D Systems™ Recombinant Mouse VSIG4 His-tag Protein CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9747VS050
Catalog No. 97-47V-S050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Mouse VSIG4 His-tag Protein is derived from NS0. The Recombinant Mouse VSIG4 His-tag Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
NP_808457
Lyophilized from a 0.2μm filtered solution in PBS.
20 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
NS0
<0.10 EU per 1μg of the protein by the LAL method.
Mouse
Bioactivity
11326
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 200μg/mL in PBS.