Learn More
Active MotifSupplier Diversity Partner Recombinant PHD1 (EGLN2) protein  DFS Item
Manufacturer:  Active MotifSupplier Diversity Partner 81064
Recombinant human PHD1 / EGLN2 protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_542770.2) with an N-terminal FLAG tag. The molecular weight of the protein is 44.9 kDa.
Catalog No. NC1482260