Learn More
Novus Biologicals™ Recombinant Porcine Flt-3 Ligand Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP2762955UG
 View more versions of this product
Catalog No. NBP2762955U
Add to cart
Description
Description
An un-tagged bioactive recombinant protein corresponding to Porcine Flt-3 Ligand
The Recombinant Porcine Flt-3 Ligand Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Lyoph
2323
>95%, by SDS-PAGE
E. coli
FLT3LG
Sterile water at 0.1 mg/mL
SDS-Page, Bioactivity
17.3 kDa
5 μg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
Flt-3 Ligand is a monomer, 17.3 kDa (155 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5