Learn More
Novus Biologicals™ Recombinant Porcine IL-2 Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP2762535UG
 View more versions of this product
Catalog No. NBP2762535U
Add to cart
Description
Description
An un-tagged bioactive recombinant protein corresponding to Porcine IL-2
The Recombinant Porcine IL-2 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Lyoph
3558
>95%, by SDS-PAGE
E. coli
IL2
Sterile water at 0.1 mg/mL
SDS-Page, Bioactivity
15.3 kDa
5 μg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
IL-2 is a monomer, 15.3 kDa (135 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5