Learn More
Novus Biologicals™ Recombinant Porcine IL-4 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP2762865UG
 View more versions of this product
Catalog No. NBP2762865U
Add to cart
Description
Description
An un-tagged bioactive recombinant protein corresponding to Porcine IL-4
The Recombinant Porcine IL-4 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Lyoph
3565
>95%, by SDS-PAGE
E. coli
IL4
Sterile water at 0.1 mg/mL
SDS-Page, Bioactivity
12.7 kDa
5 μg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
IL-4 is a monomer, 12.7 kDa (110 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5