Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Porcine IL-4 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
$131.80 - $7040.00
Specifications
Concentration | Lyoph |
---|---|
For Use With (Application) | Bioactivity, SDS-PAGE |
Gene ID (Entrez) | 3565 |
Molecular Weight (g/mol) | 12.7 kDa |
Purification Method | >95%, by SDS-PAGE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2762865U
|
Novus Biologicals™
NBP2762865UG |
5 μg |
Each for $131.80
|
|
|||||
NB27628620U
|
Novus Biologicals™
NBP27628620UG |
20 μg |
Each for $265.00
|
|
|||||
NBP2762861M
|
Novus Biologicals™
NBP2762861MG |
1 mg |
Each for $7,040.00
|
|
|||||
Description
An un-tagged bioactive recombinant protein corresponding to Porcine IL-4The Recombinant Porcine IL-4 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Lyoph | |
3565 | |
>95%, by SDS-PAGE | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IL-4 is a monomer, 12.7 kDa (110 aa) | |
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5 |
Bioactivity, SDS-PAGE | |
12.7 kDa | |
E. coli | |
IL4 | |
Sterile water at 0.1 mg/mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title