Learn More
ACROBiosystems FITC-Labeled Human Siglec-3 / CD33 Protein, His Tag
Supplier:  ACROBiosystems CD3HF22425UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human Siglec-3, His Tag (CD3-HF224) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). It is the FITC labeled form of Human Siglec-3, His Tag (CD3-H5226).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3 Antibody at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human Siglec-3, His Tag (Cat. No. CD3-HF224) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-201-9658