Learn More
R&D Systems™ Recombinant Rat GITR Ligand/TNFSF18 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9735GL050
Catalog No. 97-35GL-050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Rat GITR Ligand/TNFSF18 Protein is derived from NS0. The Recombinant Rat GITR Ligand/TNFSF18 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
XP_008767927
Lyophilized from a 0.2 μm filtered solution in PBS.
16 kDa
50 μg
12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
<0.10 EU per 1 μg of the protein by the LAL method.
GITR Ligand/TNFSF18
Reconstitute at 500 μg/mL in PBS.
Bioactivity
8995
>85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Mouse myeloma cell line, NS0-derived rat GITR Ligand/TNFSF18 protein Thr74-Ser200, with a N-terminal HA-tag
RUO
Unconjugated
Rat