Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Rat IL-2 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$818.00 - $2370.00

Specifications

Concentration 0.25 mg/mL
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity >90% SDS-PAGE
Source E. coli
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB002941
View Documents
Novus Biologicals™
NBP2774920.1MG
0.1 mg
Each for $818.00
Add to cart
 
NB002942
View Documents
Novus Biologicals™
NBP2774920.5MG
0.5 mg
Each for $2,370.00
Add to cart
 
Description

Description

Rat IL-2 stimulates cell proliferation of the CTLL2 mouse cytotoxic T cells. The ED50 range is less or equal to 0.3 ng/ml.{ATGP3889-BA.JPG}
Specifications

Specifications

0.25 mg/mL
< 1.0 EU per 1 μg of protein (determined by LAL method)
>90% SDS-PAGE
E. coli
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit