Learn More
R&D Systems™ Recombinant Rat Jagged 1 His Tagged Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9907JG050
Catalog No. 99-07JG-050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Rat Jagged 1 His Tagged Protein is derived from NS0. The Recombinant Rat Jagged 1 His Tagged Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q63722
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
114 kDa
50 μg
12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
<0.10 EU per 1 μg of the protein by the LAL method.
Jagged 1
Reconstitute at 500 μg/mL in PBS.
Bioactivity
182
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Mouse myeloma cell line, NS0-derived rat Jagged 1 protein Ser32-Asp1067, with substitutions GGARN56-60AE and DR63-64TLVRPY and a C-terminal 6-His tag
RUO
Unconjugated
Rat