Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant SARS-CoV-2 Spike RBD (N501Y) Protein
Recombinant Protein
Supplier: Novus Biologicals™ NBP307930
Specifications
Western Blot, ELISA | |
>95%, by PAGE | |
Store at -20 to -70 C. Avoid freeze-thaw cycles. | |
UK variant (B0.1.1.7) |
20 mM PBS | |
0.05 mg | |
Endotoxin level is less than 1 EU / ug by the LAL method. | |
Spike RBD |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title