Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Viral HIV-1 gp41 Protein

Recombinant Viral HIV-1 gp41 Protein

Manufacturer:  Novus Biologicals™ NBP236739

Catalog No. NBP236739

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

8M urea, 20mM Tris-HCl pH 8.0, 10mM B-mercaptoethanol
Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Viral
0.1mg
RUO - research use only
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit