Learn More
R&D Systems™ Recombinant Viral JEV-E Fc Chimera Protein CF
R&D Systems™ Recombinant Viral JEV-E Fc Chimera Protein CF is the envelope protein E produced by the Japanese encephalitis virus (JEV), a mosquito-borne flavivirus that is the sole agent of Japanese encephalitis.
Manufacturer:  R&D Systems™ 9449JV100
Catalog No. 9449JV100
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Viral JEV-E Fc Chimera Protein is derived from HEK293.
Specifications
Specifications
P32886
Bioactivity not tested
75kDa
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant Protein
Viral
Bioactivity not tested
Lyophilized from a 0.2μm filtered solution in PBS.
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
HEK293
<1.0EU per 1μg of the protein by the LAL method.
Unconjugated
Reconstitute at 500μg/mL in PBS.