Learn More
Cole-Parmer Monarch DC2C1-U12-10000 12 Channel Paperless Recorder, color, with compact flash  DFS Item
Manufacturer:  Cole-Parmer 8081310
These electronic recorders feature universal inputs for measuring DC voltage and current as well as thermocouples and RTDs for temperature measurements and display dry contact states.
Catalog No. 50-631-727