Learn More
Cole-Parmer Monarch DC2C1-U6-10000 6 Channel Paperless Recorder, color, with compact flash  DFS Item
Manufacturer:  Cole-Parmer 8081308
These electronic recorders feature universal inputs for measuring DC voltage and current as well as thermocouples and RTDs for temperature measurements and display dry contact states.
Catalog No. 50-631-735