Learn More
Recoverin Rabbit anti-Human, Unconjugated, Clone: 1J23, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB110725UL
 View more versions of this product
Catalog No. ZRB110725MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Recoverin Monoclonal antibody specifically detects Recoverin in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Recoverin
1J23
Unconjugated
Purified
RCVRN, RCV1
His-tagged recombinant full-length human Recoverin.
∼21 kDa observed; 23.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Stable for 1 year at 2 to 8°C from date of receipt.
Monoclonal
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_002894.1
Cancer-associated retinopathy protein;Protein CAR
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
Human