Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RECQL5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310393100UL
Description
RECQL5 Polyclonal specifically detects RECQL5 in Human samples. It is validated for Western Blot.Specifications
RECQL5 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
DNA helicase, RecQ-like type 5, EC 3.6.1, EC 3.6.4.12, FLJ90603, RecQ protein-like 5ATP-dependent DNA helicase Q5, RecQ5, RECQ5RecQ protein 5 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human PP1045. Peptide sequence LTPFYKEGKFASKELFKGFARHLSHLLTQKTSPGRSVKEEAQNLIRHFFH | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
9400 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction