Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Recycled Film Adhesive Warning Sign: FLAMMABLE GAS

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$15.25

Specifications

Color Black/red on white
Message FLAMMABLE GAS
Min. Temperature (English) -40°F
Quantity 1/Ea.
Mounting Type Self-adhesive
View More Specs

Products 2
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
19-104-910 View Documents Brady™
118212
14 in. x 10 in. x 0.004 in. Each for $15.25
Add to cart
 
19-104-911 View Documents Brady™
118213
10 in. x 7 in. x 0.004 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Recycled film material is abrasion-resistant and chemical-resistant
  • Bold and bright, providing high visibility to critical messages
  • For indoor use
Specifications

Specifications

Black/red on white
-40°F
Self-adhesive
7 in.
FLAMMABLE GAS
1/Ea.
10 in.
Rectangle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit