Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Recycled Film Adhesive Warning Sign: FLAMMABLE KEEP FLAMES AND HEAT AWAY

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

Manufacturer:  Brady™ 118217

 View more versions of this product

Catalog No. 19-104-913

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Recycled film material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor use
Specifications

Specifications

Safety
Not Laminated
Rectangle
Material: Recycled film
7 in.
FLAMMABLE KEEP FLAMES AND HEAT AWAY
0.004 in.
10 in.
Black/red on white
1/Ea.
10 in. x 7 in. x 0.004 in.
Self-adhesive
230°F
-40°F
Cold temperature permanent acrylic
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit