Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Recycled Film Adhesive Warning Sign: FLAMMABLE KEEP FLAMES AND HEAT AWAY

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$15.75

Specifications

Color Black/red on white
Message FLAMMABLE KEEP FLAMES AND HEAT AWAY
Min. Temperature (English) -40°F
Quantity 1/Ea.
Mounting Type Self-adhesive
View More Specs

Products 2
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
19-104-912 View Documents Brady™
118216
14 in. x 10 in. x 0.004 in. Each for $15.75
Add to cart
 
19-104-913 View Documents Brady™
118217
10 in. x 7 in. x 0.004 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Recycled film material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor use
Specifications

Specifications

Black/red on white
-40°F
Self-adhesive
7 in.
FLAMMABLE KEEP FLAMES AND HEAT AWAY
1/Ea.
10 in.
Rectangle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit