Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Recycled Film Adhesive Warning Sign: GASOLINE
Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$24.70
Specifications
Color | Black/Red on White |
---|---|
Message | GASOLINE |
Shape | Rectangular |
Temperature Range (English) | -40°F to 230°F |
Finish | Not Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-104-902
|
Brady™
118178 |
14 x 10 in. |
Each for $24.70
|
|
19-104-903
|
Brady™
118179 |
10 x 7 in. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Recycled film material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- For indoor use
Specifications
Black/Red on White | |
Rectangular | |
Not Laminated | |
1 Ea. | |
230°F | |
0.004 in. |
GASOLINE | |
-40°F to 230°F | |
Recycled film | |
Self-adhesive | |
-40°F | |
Cold temperature permanent acrylic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title