Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Recycled film Caution Sign: HAZARDOUS MATERIAL STORAGE AREA
Improve workplace safety with proper signage. The Brady™ Caution Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$24.70
Specifications
Color | Black on Yellow |
---|---|
Message | HAZARDOUS MATERIAL STORAGE AREA |
Shape | Rectangular |
Temperature Range (English) | -40°F to 230°F |
Finish | Not Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-104-916
|
Brady™
118264 |
14 x 10 in. |
Each for $24.70
|
|
19-104-917
|
Brady™
118265 |
10 x 7 in. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Excellent cold-weather performance
- Recycled film material is abrasion-resistant and chemical-resistant
- Bold and bright, providing high visibility to critical messages
Specifications
Black on Yellow | |
Rectangular | |
Not Laminated | |
1 Ea. | |
Self-adhesive | |
-40°F |
HAZARDOUS MATERIAL STORAGE AREA | |
-40°F to 230°F | |
Recycled film | |
Cold-temperature Permanent Acrylic | |
230°F | |
0.004 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title