Learn More
ZING™ Enterprises RecycLockout Gate Valve Lockout
Designed to cover and secure valve handles from being operated and releasing dangerous pressure.
Manufacturer:  ZING™ Enterprises 7104
 View more versions of this product
Catalog No. 18-002-843
Add to cart
Description
Description
Lockout is high visible red and available in 2 sizes -
Fits 1 in.-2.5 in. valves and is designed to accommodate 3 padlocks with shackles up to 3/8 in. diameter. Lockout is made from heavy-duty 100% recycled polycarbonate that is UL Validated and supports sustainability initiatives and LEED credits. Lockout measures 1 in.H x 3 in. diameter. Made in the USA.
Fits 2.5 in. - 5 in. valves and is designed to accommodate 3 padlocks with shackles up to 3/8 in. diameter. Lockout is made from heavy-duty 100% recycled polycarbonate that is UL Validated and supports sustainability initiatives and LEED credits. Lockout measures 1-3/4 in.H x 5-3/4 in. diameter. Made in the USA.
Specifications
Specifications
Lockout
5.75 in.
Safety
1
5.75 in.
Red
English
5.75 in.
5.75 in. x 5.75 in.
Plastic