Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZING™ Enterprises RecycLockout Lockout Tagout, Wall Switch Lockout, Recycled Plastic

Wall Switch Lockout ensures safety with a compact design.

Manufacturer:  ZING™ Enterprises 6064

Catalog No. 18-002-966


Add to cart

Description

Description

Lockout fits easily over a wide variety of wall switches and locks the switch in either the  in.on in. or  in.off in. position. Lockout is high visible red and will accommodate 1 padlock with shackles up to 3/8 in. diameter. Wall switch lockout is constructed from heavy-duty 100% recycled polypropylene that is UL Validated and supports sustainability initiatives and LEED credits. Switch lockout measures 3-1/2 in.H x 1-1/2 in. W x 1/4 in.D. Made in the USA.

Specifications

Specifications

1
Safety
3.5 in.
Plastic
3.5 in. x 1.5 in.
Red
Lockout
Floor standing
1.5 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit