Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VF Workwear Red Kap Men's Lab Coats

With four-button closure

Manufacturer:  VF Workwear 5700WH LN L

 View more versions of this product

Catalog No. 19-166-276


Add to cart

Description

Description

  • 80% polyester/20% cotton blend
  • Pre-cure durable press finish
  • One piece, lined, notched lapel collar
  • Left breast pocket, bartacked pencil stall, two lower pockets
  • Side-vent openings
  • White (navy blue available in long, medium size)

Specifications

Specifications

Notched lapel
White
3
Pre-cure durable press
Button Front
Long
41.5 in.
Button
Cotton, Polyester
Large
Male
44 to 46 in.
105.4cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit