Red Kap™ Unisex Specialized Cuff Lab Coats
Provides protection in many work environments
 View more versions of this product
Catalog No. 19-166-232
  Check Availability
Add to Cart
    Specifications
    Color White
    Material Polyester, Cotton
    No. of Pockets 3.0
    Size 2X-Large
    Finish Pre-cure durable press
    Gender Unisex
    Collar Type Notched lapel
    Style Long
    Closure Type Gripper
    Length (English) 41.5 in.
    Construction Polyester, Combed Cotton
    Cuff Style Knit
    Length (Metric) 105.41cm
  • Description & Specifications
Related Products
Product Content Feedback