Learn More
Cole-Parmer Masterflex Fitting, PVDF, Straight, Hose Barb Reducer, 3/8" ID x 1/4" ID; 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3061620
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation. Select the fitting based on the Masterflex tubing size. Fittings are available in a variety of materials to suit different applications and budgets.
Catalog No. 50-629-522