Learn More
Cole-Parmer Masterflex Fitting, Polyethylene, Straight, Threaded Reducer, 3/8" NPT(M) x 1/2" NPT(M); 100/PK  DFS Item
Manufacturer:  Cole-Parmer 3062752
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
Catalog No. 50-630-249