Learn More
Cole-Parmer Masterflex Fitting, PVDF, Straight, Threaded Reducer, 1/8" NPT(M) x 1/2" NPT(M); 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3062345
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
Catalog No. 50-630-229