Red Rubber Sleeve Stoppers for Serum and Vaccine Bottles

Manufacturer: PlasticoidSupplier Diversity Partner 20220

 View more versions of this product

Catalog No. 14-126BB

  Check Availability
Add to Cart

    Specifications

    Diameter (English) Plug 0.54 in.
    Length (English) Sleeve 0.62 in.
    Length (Metric) Sleeve 1.5cm
    Material Rubber
    Diameter (Metric) Plug 1.38cm
    For Use With (Equipment) Serum and vaccine bottles
    Size 14.5
    Stopper Number 14.5
    Color Red
    Product Type Sleeve Stopper

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit