Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red fluorescent protein eqFP611 Rabbit anti-All Species, Clone: JA10-52, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP267102

Catalog No. NBP267102

Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.


Description

Description

Red fluorescent protein eqFP611 Monoclonal specifically detects Red fluorescent protein eqFP611 in All Species samples. It is validated for Western Blot.
Specifications

Specifications

RedfluorescentproteineqFP611
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000
GFP-like chromoprotein, Red fluorescent protein eqFP611
Recombinant protein within Entacmaea quadricolor Red fluorescent protein eqFP611 RFP aa 1-216/ 231. (SwissProt: Q8ISF8 EntacmaeaQuadricolorEqFP611)
100 μL
All species
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
JA10-52
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Rabbit
Protein A purified
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit