Learn More
Sklar InstrumentsSupplier Diversity Partner Redon Spike, Sklar, 7-1/2 in. Redon Spike 8 French, Triangular Tip, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 261920
Redon Spike, Sklar, 7-1/2 in. Redon Spike 8 French, Triangular Tip, Stainless Steel. Sklar's Redon Spike is a needle/trocar most commonly used as part of a Redon wound drainage set, which are frequently utilized following hernia procedures. The spike, available with various tip styles in various sizes, is used to insert tubing near the surgical site, which then connects to a drainage container. Fluid is drained from the surgical site following the procedure so that it does not collect at the surgical site and result in hemostasis or other complications. This product is slightly curved with a French 8, triangular tip, and overall length of 7-1/2 inches.
Catalog No. 50-143-8055