Learn More
Cole-Parmer Masterflex Fitting, HDPE, Tee, Hose Barb to Threaded Adapter, 3/8" ID x 3/8" ID x 1/4" NTP(M); 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3062406
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
Catalog No. 50-630-575