Learn More
Reduced Folate Carrier/SLC19A1 Mouse anti-Human, Alexa Fluor 405, Clone: 890513, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8450V100UG
Catalog No. FAB8450V100
Add to cart
This item is not returnable. View return policy
Description
Description
Reduced Folate Carrier/SLC19A1 Monoclonal antibody specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Reduced Folate Carrier/SLC19A1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P41440
SLC19A1
NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1 Met1-Gln591 Accession # P41440
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains human Reduced Folate Carrier/SLC19A1 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
890513
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CHMD, FLOT1, folate transporter 1, FOLT, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1
Mouse
IgG2a
RUO
Primary
6573
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)