Learn More
Cole-Parmer Parker Hannifin PNR-84-P Threaded Reducing Nipples, PFA, 1/2 x 1/4" NPT(M); 1/Ea  DFS Item
Manufacturer:  Cole-Parmer 3132103
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.
Catalog No. 50-632-036