Learn More
Cole-Parmer Masterflex Fitting, HDPE, Tee, Hose Barb Reducer, 5/8" ID x 5/8" ID x 3/8" ID; 100/PK  DFS Item
Manufacturer:  Cole-Parmer 3040162
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-630-260